Product Details

SNP ID
rs1557873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:25343215 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACATAAACAAGAAAAACAGAATT[C/T]GGAAGTCTAAATAAAGTTATTATGG
Phenotype
MIM: 601623
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
UBE3A PubMed Links

Gene Details

Gene
UBE3A
Gene Name
ubiquitin protein ligase E3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000462.3 Intron NP_000453.2
NM_130838.1 Intron NP_570853.1
NM_130839.2 Intron NP_570854.1
XM_005268267.4 Intron XP_005268324.1
XM_005268268.4 Intron XP_005268325.1
XM_005268269.4 Intron XP_005268326.1
XM_005268270.4 Intron XP_005268327.1
XM_005268271.4 Intron XP_005268328.1
XM_006720675.3 Intron XP_006720738.1
XM_006720676.3 Intron XP_006720739.1
XM_011521994.2 Intron XP_011520296.1
XM_011521995.2 Intron XP_011520297.1
XM_017022544.1 Intron XP_016878033.1
XM_017022545.1 Intron XP_016878034.1
XM_017022546.1 Intron XP_016878035.1
XM_017022547.1 Intron XP_016878036.1
XM_017022548.1 Intron XP_016878037.1
XM_017022549.1 Intron XP_016878038.1
XM_017022550.1 Intron XP_016878039.1
XM_017022551.1 Intron XP_016878040.1
XM_017022552.1 Intron XP_016878041.1
XM_017022553.1 Intron XP_016878042.1
XM_017022554.1 Intron XP_016878043.1
XM_017022555.1 Intron XP_016878044.1
XM_017022556.1 Intron XP_016878045.1
XM_017022557.1 Intron XP_016878046.1
XM_017022558.1 Intron XP_016878047.1
XM_017022559.1 Intron XP_016878048.1

View Full Product Details