Product Details

SNP ID
rs10043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:56106422 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGGCTTCCACGCGGCATCCTAGGTA[A/C]GCGACGGCTAGGCCTCCGGACGTGG
Phenotype
MIM: 616244
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHCHD2 PubMed Links

Gene Details

Gene
CHCHD2
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320327.1 Intron NP_001307256.1
NM_016139.3 Intron NP_057223.1
Gene
NUPR2
Gene Name
nuclear protein 2, transcriptional regulator
There are no transcripts associated with this gene.

View Full Product Details