Product Details

SNP ID
rs944953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124859726 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCTGTGCACACCTCCCAGAACACT[A/C]AAGCCGACCCTCCCCAAGGTAAGGT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
There are no transcripts associated with this gene.

Gene
RPL35
Gene Name
ribosomal protein L35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007209.3 Intron NP_009140.1
Gene
WDR38
Gene Name
WD repeat domain 38
There are no transcripts associated with this gene.

View Full Product Details