Product Details

SNP ID
rs1217339
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:69432683 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTCGCATGAGGCTGCAGATCTGCCA[A/G]AGTCAAAGGCAGAGTTACCCTCATT
Phenotype
MIM: 602414
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APBA1 PubMed Links

Gene Details

Gene
APBA1
Gene Name
amyloid beta precursor protein binding family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163.3 Intron NP_001154.2
XM_005251968.3 Intron XP_005252025.1
XM_011518617.2 Intron XP_011516919.1
XM_017014670.1 Intron XP_016870159.1

View Full Product Details