Product Details

SNP ID
rs1217338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:69433182 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGAGGGGAGAGACTCCTTCAGAG[C/T]TTCCCAATTCAGCGGCCCCTGTTCA
Phenotype
MIM: 602414
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APBA1 PubMed Links

Gene Details

Gene
APBA1
Gene Name
amyloid beta precursor protein binding family A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163.3 Intron NP_001154.2
XM_005251968.3 Intron XP_005252025.1
XM_011518617.2 Intron XP_011516919.1
XM_017014670.1 Intron XP_016870159.1

View Full Product Details