Product Details

SNP ID
rs1747837
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136114722 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTGCAGCGTTGACACCGCACGA[A/G]CGGCACCACCAGCCCCCTCCCGCCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C9orf69 PubMed Links
Additional Information
For this assay, SNP(s) [rs111715438] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C9orf69
Gene Name
chromosome 9 open reading frame 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256526.1 2663 UTR 3 NP_001243455.1
NM_152833.2 2663 UTR 3 NP_690046.3
Gene
LOC105376323
Gene Name
uncharacterized LOC105376323
There are no transcripts associated with this gene.

Gene
LOC107987142
Gene Name
translation initiation factor IF-2-like
There are no transcripts associated with this gene.

View Full Product Details