Product Details
- SNP ID
-
rs1536857
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:474053 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTAATGAAGAAGCTTTCCCCACA[A/T]GATGGCTCCTCACAAGAAATACTAT
- Phenotype
-
MIM: 611432
MIM: 607704
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DOCK8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150886772] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DOCK8
- Gene Name
- dedicator of cytokinesis 8
There are no transcripts associated with this gene.
- Gene
- KANK1
- Gene Name
- KN motif and ankyrin repeat domains 1
View Full Product Details