Product Details
- SNP ID
-
rs1441724
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:163526664 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTAAGGTCCACATGTTTTTTAAAA[C/T]GTTAATTACTTATATCCTTATTTAT
- Phenotype
-
MIM: 613331
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MARCH1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149047048] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MARCH1
- Gene Name
- membrane associated ring-CH-type finger 1
- Gene
- TMA16
- Gene Name
- translation machinery associated 16 homolog
There are no transcripts associated with this gene.
View Full Product Details