Product Details

SNP ID
rs1542185
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27239117 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGTGCTGGCCAGCTCCAGGGCCTA[C/T]GCAACATTCAGACACCTTGTGAAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STMN4 PubMed Links

Gene Details

Gene
STMN4
Gene Name
stathmin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283053.1 Intron NP_001269982.1
NM_001283054.1 Intron NP_001269983.1
NM_001283055.1 Intron NP_001269984.1
NM_030795.3 Intron NP_110422.2
XM_005273652.4 Intron XP_005273709.1
XM_005273655.4 Intron XP_005273712.1

View Full Product Details