Product Details

SNP ID
rs1442794
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:49331273 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAAGAAGCCCTTCAGACTTAACA[A/C]GAATGCCCATAAGTAAAGAGAAGCA
Phenotype
MIM: 137028
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
FAM227B PubMed Links

Gene Details

Gene
FAM227B
Gene Name
family with sequence similarity 227 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152647.2 Intron NP_689860.2
Gene
GALK2
Gene Name
galactokinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001556.2 Intron NP_001001556.1
NM_001289030.1 Intron NP_001275959.1
NM_001289031.1 Intron NP_001275960.1
NM_002044.3 Intron NP_002035.1

View Full Product Details