Product Details

SNP ID
rs1530836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41982338 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCTGAAGGGAGACCTGCTGACTT[G/T]GGGTTTCTCTGTGTCCCAGGGTTTT
Phenotype
MIM: 605892
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EHD4 PubMed Links

Gene Details

Gene
EHD4
Gene Name
EH domain containing 4
There are no transcripts associated with this gene.

Gene
PLA2G4E
Gene Name
phospholipase A2 group IVE
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206670.1 3933 UTR 3 NP_001193599.1
XM_011521237.2 3933 UTR 3 XP_011519539.1
XM_011521238.1 3933 Intron XP_011519540.1
XM_011521239.1 3933 Intron XP_011519541.1
Gene
PLA2G4E-AS1
Gene Name
PLA2G4E antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details