Product Details

SNP ID
rs1044219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42159006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGCTATCCAGGGAGGTGGCCCA[A/G]AGCTTCCCTGCTGCTCCTGGAGGAA
Phenotype
MIM: 612188
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLA2G4F PubMed Links

Gene Details

Gene
PLA2G4F
Gene Name
phospholipase A2 group IVF
There are no transcripts associated with this gene.

Gene
VPS39
Gene Name
VPS39, HOPS complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301138.1 4621 UTR 3 NP_001288067.1
NM_015289.3 4621 UTR 3 NP_056104.2
XM_011521403.2 4621 UTR 3 XP_011519705.1
XM_011521404.2 4621 UTR 3 XP_011519706.1
XM_017022036.1 4621 Intron XP_016877525.1
XM_017022037.1 4621 Intron XP_016877526.1
XM_017022038.1 4621 UTR 3 XP_016877527.1
XM_017022039.1 4621 UTR 3 XP_016877528.1

View Full Product Details