Product Details
- SNP ID
-
rs1703755
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:42212978 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCTACAGACACACTGATTCAATAA[C/T]ACATGGAAAAATTCCCTTTGTGGGA
- Phenotype
-
MIM: 612188
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM87A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115137148] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TMEM87A
- Gene Name
- transmembrane protein 87A
- Gene
- VPS39
- Gene Name
- VPS39, HOPS complex subunit
There are no transcripts associated with this gene.
View Full Product Details