Product Details
- SNP ID
-
rs1050666
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:108622992 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCCGAGGAAGCCCAGAGCTGTGGA[A/T]TGGGGTCTGGGCACTCAGGGGTGGC
- Phenotype
-
MIM: 600738
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
SELPLG
PubMed Links
Gene Details
- Gene
- SELPLG
- Gene Name
- selectin P ligand
View Full Product Details