Product Details

SNP ID
rs1444193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:58639904 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTAAATAAAGCATTGCACTTGAAT[C/T]CCTGTCTCAGGATCTGAGTCTGGGA
Phenotype
MIM: 608619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FAM3D PubMed Links

Gene Details

Gene
FAM3D
Gene Name
family with sequence similarity 3 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138805.2 Intron NP_620160.1
XM_005264862.4 Intron XP_005264919.1
XM_005264863.4 Intron XP_005264920.1
XM_005264864.4 Intron XP_005264921.1
XM_006712965.3 Intron XP_006713028.1
XM_006712967.3 Intron XP_006713030.1
XM_011533348.2 Intron XP_011531650.1
XM_011533349.2 Intron XP_011531651.1
XM_011533350.2 Intron XP_011531652.1
XM_011533351.2 Intron XP_011531653.1
XM_011533352.2 Intron XP_011531654.1
XM_011533353.2 Intron XP_011531655.1
XM_011533354.2 Intron XP_011531656.1
Gene
FAM3D-AS1
Gene Name
FAM3D antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details