Product Details

SNP ID
rs1063611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:41744145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGGTACACTCTTGGAAAACTATA[A/T]GCACATGTAGAAATATTTCCCTTTT
Phenotype
MIM: 603851
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PHOX2B PubMed Links

Gene Details

Gene
PHOX2B
Gene Name
paired like homeobox 2b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003924.3 2967 UTR 3 NP_003915.2

View Full Product Details