Product Details

SNP ID
rs953756
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:219973073 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGTGGCACTTAGAAAAATAACC[A/G]TAATACAAGTAAAAAACCCAGAGAG
Phenotype
MIM: 138295
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EPRS PubMed Links

Gene Details

Gene
EPRS
Gene Name
glutamyl-prolyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004446.2 Intron NP_004437.2
XM_017000614.1 Intron XP_016856103.1

View Full Product Details