Product Details

SNP ID
rs1046927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:37250862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGCAATTACTTCTTTTACAAACA[C/T]GACTTAAAGACAAAAAGTTATGCAG
Phenotype
MIM: 615898 MIM: 607077
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFAF7 PubMed Links

Gene Details

Gene
NDUFAF7
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083946.1 5821 Intron NP_001077415.1
NM_144736.4 5821 Intron NP_653337.1
XM_005264410.1 5821 Intron XP_005264467.1
XM_005264411.1 5821 Intron XP_005264468.1
XM_011532956.1 5821 Intron XP_011531258.1
Gene
PRKD3
Gene Name
protein kinase D3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005813.4 5821 UTR 3 NP_005804.1
XM_005264237.3 5821 UTR 3 XP_005264294.1
XM_005264238.3 5821 UTR 3 XP_005264295.1
XM_017003741.1 5821 UTR 3 XP_016859230.1
XM_017003742.1 5821 UTR 3 XP_016859231.1
XM_017003743.1 5821 UTR 3 XP_016859232.1
XM_017003744.1 5821 UTR 3 XP_016859233.1

View Full Product Details