Product Details

SNP ID
rs1199446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70305074 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTATGAACTTCTGTGTGTAAATT[C/T]TCACGTGGACAAAGGTTTTCATTTA
Phenotype
MIM: 300521
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF4A PubMed Links

Gene Details

Gene
KIF4A
Gene Name
kinesin family member 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012310.4 Intron NP_036442.3

View Full Product Details