Product Details

SNP ID
rs1540600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:17219170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTATACTGCAGGCAAAATGAAT[A/G]AATCGGGACTGTAATTACCTATTCC
Phenotype
MIM: 605940
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BASP1 PubMed Links

Gene Details

Gene
BASP1
Gene Name
brain abundant membrane attached signal protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271606.1 Intron NP_001258535.1
NM_006317.4 Intron NP_006308.3
Gene
LOC285696
Gene Name
uncharacterized LOC285696
There are no transcripts associated with this gene.

View Full Product Details