Product Details

SNP ID
rs1062712
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:138631685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGACAGAAAACTGCCTGTACAT[A/G]CTATGTCCACTTTTGGAACACAGAT
Phenotype
MIM: 300070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF13 PubMed Links

Gene Details

Gene
FGF13
Gene Name
fibroblast growth factor 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139498.1 2403 UTR 3 NP_001132970.1
NM_001139500.1 2403 UTR 3 NP_001132972.1
NM_001139501.1 2403 UTR 3 NP_001132973.1
NM_001139502.1 2403 UTR 3 NP_001132974.1
NM_004114.3 2403 UTR 3 NP_004105.1
NM_033642.2 2403 UTR 3 NP_378668.1
XM_005262399.1 2403 UTR 3 XP_005262456.1

View Full Product Details