Product Details

SNP ID
rs1138276
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.4:88091827 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AGTATAGCATTAATACATTTGTCAT[A/T]TATTAAATAATGGCAAACCATAGCA
Phenotype
MIM: 603756
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ABCG2 PubMed Links

Gene Details

Gene
ABCG2
Gene Name
ATP binding cassette subfamily G member 2 (Junior blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257386.1 2982 UTR 3 NP_001244315.1
NM_004827.2 2982 UTR 3 NP_004818.2
XM_005263354.3 2982 UTR 3 XP_005263411.1
XM_005263355.3 2982 UTR 3 XP_005263412.1
XM_005263356.3 2982 UTR 3 XP_005263413.1
XM_011532420.2 2982 UTR 3 XP_011530722.1
XM_017008852.1 2982 UTR 3 XP_016864341.1
XM_017008853.1 2982 UTR 3 XP_016864342.1

View Full Product Details