Product Details

SNP ID
rs2018417
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.4:99313983 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CAAATTTGTCCTTGTTGATGTCCAC[A/C]GCAATGATTCTGGCTGCTCCAGCTG
Phenotype
MIM: 103720
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ADH1B PubMed Links

Gene Details

Gene
ADH1B
Gene Name
alcohol dehydrogenase 1B (class I), beta polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000668.5 899 Silent Mutation GCG,GCT A222A NP_000659.2
NM_001286650.1 899 Silent Mutation GCG,GCT A182A NP_001273579.1

View Full Product Details