Product Details
- SNP ID
-
rs919804
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48365921 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTGAGAAGGCCCAAGACCATCACG[C/T]GGGTCTTCGAAGGGGTGAGGCCCTC
- Phenotype
-
MIM: 608373
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SYNGR4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76242262] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SYNGR4
- Gene Name
- synaptogyrin 4
- Gene
- TMEM143
- Gene Name
- transmembrane protein 143
There are no transcripts associated with this gene.
View Full Product Details