Product Details

SNP ID
rs873171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64539844 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGAGGAGTGAGTGGGAAGTTAG[A/G]AAGTCGAGAGAGTGAATAGGCACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AFTPH PubMed Links

Gene Details

Gene
AFTPH
Gene Name
aftiphilin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002243.2 Intron NP_001002243.1
NM_017657.4 Intron NP_060127.3
NM_203437.3 Intron NP_982261.2
XM_005264380.1 Intron XP_005264437.1

View Full Product Details