Product Details

SNP ID
rs1011369
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:92901596 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTTTTTCTGTCAGTAAATTTCA[A/G]TTTTTAGGTTTTTGTACTGCAGAAA
Phenotype
MIM: 602119
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHD2 PubMed Links

Gene Details

Gene
CHD2
Gene Name
chromodomain helicase DNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042572.2 Intron NP_001036037.1
NM_001271.3 Intron NP_001262.3
Gene
LINC01578
Gene Name
long intergenic non-protein coding RNA 1578
There are no transcripts associated with this gene.

Gene
MIR3175
Gene Name
microRNA 3175
There are no transcripts associated with this gene.

View Full Product Details