Product Details

SNP ID
rs817340
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63974671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAATCCAGGCCTTCTTGGGAAAG[A/C]AGCAATGGGAAGGCCCACGCACAAG
Phenotype
MIM: 613979
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRPF6 PubMed Links

Gene Details

Gene
PRPF6
Gene Name
pre-mRNA processing factor 6
There are no transcripts associated with this gene.

Gene
SAMD10
Gene Name
sterile alpha motif domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080621.4 1623 UTR 3 NP_542188.1
XM_005260199.4 1623 UTR 3 XP_005260256.1
XM_006723705.3 1623 UTR 3 XP_006723768.1
XM_011528565.2 1623 UTR 3 XP_011526867.1
XM_017027671.1 1623 UTR 3 XP_016883160.1
Gene
ZNF512B
Gene Name
zinc finger protein 512B
There are no transcripts associated with this gene.

View Full Product Details