Product Details
- SNP ID
-
rs1511025
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:119319083 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TCCTTTTGGCTTCTACTCCTTCATA[C/T]ACATAAGTCTGAAAGGTGTTAACAA
- Phenotype
-
MIM: 134640
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FABP2
PubMed Links
Gene Details
- Gene
- FABP2
- Gene Name
- fatty acid binding protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000134.3 |
416 |
Silent Mutation |
GTA,GTG |
V119V |
NP_000125.2 |
View Full Product Details