Product Details

SNP ID
rs1497566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:46038599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGTGGTGAAACTTATACTTGGAA[G/T]AGAGGAACAGGAAATGAACTGAATC
Phenotype
MIM: 137166
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GABRG1 PubMed Links

Gene Details

Gene
GABRG1
Gene Name
gamma-aminobutyric acid type A receptor gamma1 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173536.3 3939 UTR 3 NP_775807.2
XM_017007990.1 3939 UTR 3 XP_016863479.1

View Full Product Details