Product Details

SNP ID
rs1060459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157744152 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCAGGAGCGGGCCGGAGGGGTCC[G/T]GGTCGGAGCAGGAAGGCGCCACGCA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LSM11 PubMed Links

Gene Details

Gene
LSM11
Gene Name
LSM11, U7 small nuclear RNA associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173491.3 458 Silent Mutation CCG,CCT P134P NP_775762.1
Gene
THG1L
Gene Name
tRNA-histidine guanylyltransferase 1 like
There are no transcripts associated with this gene.

View Full Product Details