Product Details

SNP ID
rs1000934
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218826799 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAAGTCTCAGAGTAGACGGAGAC[G/T]TCTAATCCCCAGTCCATCCTGTACC
Phenotype
MIM: 604976
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MIR9500 PubMed Links
Additional Information
For this assay, SNP(s) [rs76235829] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
MIR9500
Gene Name
microRNA 9500
There are no transcripts associated with this gene.

Gene
PRKAG3
Gene Name
protein kinase AMP-activated non-catalytic subunit gamma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017431.2 Intron NP_059127.2
XM_017004343.1 Intron XP_016859832.1

View Full Product Details