Product Details
- SNP ID
-
rs1296224
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:158703335 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCTCTGCATTTTTCTCTTTAAATCC[A/C]AAATATAGTTGCATTTTCCAAAGTT
- Phenotype
-
MIM: 164343
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EBF1
PubMed Links
Gene Details
- Gene
- EBF1
- Gene Name
- early B-cell factor 1
View Full Product Details