Product Details
- SNP ID
-
rs1800971
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:6870594 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GATTCATTTTGGCCCAAAGGAGGTA[C/T]ACTTCAAAAGCAATTTTCCTTTATT
- Phenotype
-
MIM: 602056
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5743491] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DEFB1
- Gene Name
- defensin beta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005218.3 |
444 |
UTR 3 |
|
|
NP_005209.1 |
View Full Product Details