Product Details

SNP ID
rs1109138
Assay Type
Functionally tested
NCBI dbSNP Submissions
59
Location
Chr.1:109735327 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAGCTGGGACTATGCATGAGCAA[C/T]TGCGCACAGCTAATTAAAAAAAATT
Phenotype
MIM: 138390
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTM3 PubMed Links

Gene Details

Gene
GSTM3
Gene Name
glutathione S-transferase mu 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000849.4 2732 UTR 3 NP_000840.2

View Full Product Details