Product Details

SNP ID
rs1027463
Assay Type
Functionally Tested
NCBI dbSNP Submissions
25
Location
Chr.1:110060421 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCAAGTAAGCTGTCTTAGCTGGA[C/G]AGATGACAATGGACTTGGACAAAGT
Phenotype
MIM: 606014
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ALX3 PubMed Links

Gene Details

Gene
ALX3
Gene Name
ALX homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006492.2 1432 UTR 3 NP_006483.2
Gene
STRIP1
Gene Name
striatin interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details