Product Details

SNP ID
rs884176
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:106276121 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGATGTGGTTCCTTCTGATTTTGT[C/G]ATTCAGAATTGTCTCTATCCTCCTA
Phenotype
MIM: 605609
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OXR1 PubMed Links

Gene Details

Gene
OXR1
Gene Name
oxidation resistance 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198532.1 Intron NP_001185461.1
NM_001198533.1 Intron NP_001185462.1
NM_001198534.1 Intron NP_001185463.1
NM_001198535.1 Intron NP_001185464.1
NM_018002.3 Intron NP_060472.2
NM_181354.4 Intron NP_851999.2
XM_006716595.2 Intron XP_006716658.1
XM_017013589.1 Intron XP_016869078.1
XM_017013590.1 Intron XP_016869079.1
XM_017013591.1 Intron XP_016869080.1
XM_017013592.1 Intron XP_016869081.1
XM_017013593.1 Intron XP_016869082.1
XM_017013594.1 Intron XP_016869083.1
XM_017013595.1 Intron XP_016869084.1
XM_017013596.1 Intron XP_016869085.1
XM_017013597.1 Intron XP_016869086.1
XM_017013598.1 Intron XP_016869087.1
XM_017013599.1 Intron XP_016869088.1
XM_017013600.1 Intron XP_016869089.1

View Full Product Details