Product Details

SNP ID
rs1153940
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:223783617 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGACCATACGCCAGCCTTCCC[C/T]TGAAAATACTCCAGGAGAATCTTTA
Phenotype
MIM: 114230 MIM: 602143
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CAPN2 PubMed Links

Gene Details

Gene
CAPN2
Gene Name
calpain 2
There are no transcripts associated with this gene.

Gene
TP53BP2
Gene Name
tumor protein p53 binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031685.2 Intron NP_001026855.2
NM_005426.2 Intron NP_005417.1
XM_011544268.1 Intron XP_011542570.1
XM_011544269.2 Intron XP_011542571.1
XM_017002220.1 Intron XP_016857709.1

View Full Product Details