Product Details
- SNP ID
-
rs1047597
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:15835559 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCTCTAGGTTTCTCATGGCTACCC[C/G]CTGCCCTCTCCCCGTGCTGGGGAGT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ37453
PubMed Links
Gene Details
- Gene
- FLJ37453
- Gene Name
- uncharacterized LOC729614
There are no transcripts associated with this gene.
View Full Product Details