Product Details
- SNP ID
-
rs1052596
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:15492351 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAACAATTTTTAAAATTTCACTTA[A/G]AATTCATAGATTACAGCTGTTCAGA
- Phenotype
-
MIM: 602234
MIM: 609444
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CASP9
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75093591] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CASP9
- Gene Name
- caspase 9
- Gene
- CELA2B
- Gene Name
- chymotrypsin like elastase family member 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015849.2 |
1638 |
Intron |
|
|
NP_056933.2 |
View Full Product Details