Product Details
- SNP ID
-
rs1048260
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
37
- Location
-
Chr.1:182573716 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCAGGTGCTCATTACAAATCAGA[C/G]AACCAAAGCTTCATTTTAAAAGTTA
- Phenotype
-
MIM: 180435
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEL
PubMed Links
Gene Details
- Gene
- RNASEL
- Gene Name
- ribonuclease L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021133.3 |
4159 |
UTR 3 |
|
|
NP_066956.1 |
View Full Product Details