Product Details

SNP ID
rs3847357
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:70153660 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAAATACCTGATGAATGAATCCC[A/G]TTTAAAACGTTAAAAAGAACACACT
Phenotype
MIM: 607691 MIM: 611017
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SAR1A PubMed Links

Gene Details

Gene
SAR1A
Gene Name
secretion associated Ras related GTPase 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142648.1 Intron NP_001136120.1
NM_020150.4 Intron NP_064535.1
Gene
TYSND1
Gene Name
trypsin domain containing 1
There are no transcripts associated with this gene.

View Full Product Details