Product Details
- SNP ID
-
rs1517602
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
26
- Location
-
Chr.1:84555874 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGGGGAATCAATGCTCTGGTCCTCT[A/G]AAACCAGCTTATGTGCTTTTTTGTA
- Phenotype
-
MIM: 600873
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CTBS
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs72950335] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CTBS
- Gene Name
- chitobiase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004388.2 |
|
Intron |
|
|
NP_004379.1 |
- Gene
- SPATA1
- Gene Name
- spermatogenesis associated 1
View Full Product Details