Product Details

SNP ID
rs1517904
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:86645488 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATGGAGATTGCTAGTCTTATCCAGG[A/C]CATTGATTAATACTACATCTCAACT
Phenotype
MIM: 604337
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CLCA3P PubMed Links

Gene Details

Gene
CLCA3P
Gene Name
chloride channel accessory 3, pseudogene
There are no transcripts associated with this gene.

Gene
LOC105378828
Gene Name
uncharacterized LOC105378828
There are no transcripts associated with this gene.

View Full Product Details