Product Details

SNP ID
rs1497622
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:143577615 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTTTTCCCTCTTAAAGTCTTTCAA[C/T]CAAAGGAACTACAAGCACTCTGCTT
Phenotype
MIM: 608946
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FREM3 PubMed Links

Gene Details

Gene
FREM3
Gene Name
FRAS1 related extracellular matrix 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168235.1 6416 Missense Mutation GAT,GGT D2139G NP_001161707.1

View Full Product Details