Product Details

SNP ID
rs1519406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:144974834 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGACTCAATATTGTAAAAGTATCA[C/T]TTTTCCTCAGATTGATCTATAGTAT
Phenotype
MIM: 613745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANAPC10 PubMed Links

Gene Details

Gene
ANAPC10
Gene Name
anaphase promoting complex subunit 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256706.1 Intron NP_001243635.1
NM_001256707.1 Intron NP_001243636.1
NM_001256708.1 Intron NP_001243637.1
NM_001256709.1 Intron NP_001243638.1
NM_001256710.1 Intron NP_001243639.1
NM_001256711.1 Intron NP_001243640.1
NM_001256712.1 Intron NP_001243641.1
NM_001318367.1 Intron NP_001305296.1
NM_014885.4 Intron NP_055700.2
XM_011531524.2 Intron XP_011529826.1
XM_011531525.2 Intron XP_011529827.1
XM_011531526.2 Intron XP_011529828.1
XM_011531527.2 Intron XP_011529829.1
XM_011531529.2 Intron XP_011529831.1
XM_011531530.2 Intron XP_011529832.1
XM_011531531.2 Intron XP_011529833.1
XM_011531532.2 Intron XP_011529834.1
XM_017007638.1 Intron XP_016863127.1
XM_017007639.1 Intron XP_016863128.1
XM_017007640.1 Intron XP_016863129.1
XM_017007641.1 Intron XP_016863130.1
XM_017007642.1 Intron XP_016863131.1
XM_017007643.1 Intron XP_016863132.1
XM_017007644.1 Intron XP_016863133.1

View Full Product Details