Product Details

SNP ID
rs1127648
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75674754 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGACCCTCGATGAGCCCCAGCCTA[A/G]CCTGCTCCAAAGCCACTGACACATG
Phenotype
MIM: 601172
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CSPG4 PubMed Links

Gene Details

Gene
CSPG4
Gene Name
chondroitin sulfate proteoglycan 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001897.4 7858 UTR 3 NP_001888.2

View Full Product Details