Product Details
- SNP ID
-
rs1532155
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:75203986 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTGAGGTGTGGGGTTCTGTCCAGGA[A/G]ATGAACCAGCTTGCTTTCTGGGTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C15orf39
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs201909106] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C15orf39
- Gene Name
- chromosome 15 open reading frame 39
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015492.4 |
|
Intron |
|
|
NP_056307.2 |
View Full Product Details