Product Details

SNP ID
rs1799634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75848031 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAAATGTTGGAGCTGTGAATCTA[G/T]GAGGAAGTGAGAAGTTTAGCTTTTA
Phenotype
MIM: 612501
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
UBE2Q2 PubMed Links

Gene Details

Gene
UBE2Q2
Gene Name
ubiquitin conjugating enzyme E2 Q2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145335.1 Intron NP_001138807.1
NM_001284382.1 Intron NP_001271311.1
NM_173469.3 Intron NP_775740.1
XM_005254787.1 Intron XP_005254844.1
XM_005254788.1 Intron XP_005254845.1
XM_006720770.1 Intron XP_006720833.1
XM_011522228.1 Intron XP_011520530.1
XM_017022726.1 Intron XP_016878215.1
XM_017022727.1 Intron XP_016878216.1
XM_017022728.1 Intron XP_016878217.1

View Full Product Details