Product Details

SNP ID
rs1294351
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141986859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATACCTCCTAGGGTATGTGTACCCT[C/G]ATCTAAAAAAATAGAACTCAATGGG
Phenotype
MIM: 154360
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MGAM PubMed Links

Gene Details

Gene
MGAM
Gene Name
maltase-glucoamylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004668.2 Intron NP_004659.2
XM_006716168.2 Intron XP_006716231.1
XM_011516670.2 Intron XP_011514972.1
XM_011516671.2 Intron XP_011514973.1
XM_011516672.2 Intron XP_011514974.1
XM_011516673.2 Intron XP_011514975.1
XM_011516674.2 Intron XP_011514976.1
XM_017012772.1 Intron XP_016868261.1

View Full Product Details