Product Details

SNP ID
rs1803255
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:207886707 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCAACAGCAAACAAGGACCTGG[A/G]TCACCCACAGAAGAGGCAGCTGGTG
Phenotype
MIM: 142230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD34 PubMed Links

Gene Details

Gene
CD34
Gene Name
CD34 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025109.1 2642 UTR 3 NP_001020280.1
NM_001773.2 2642 UTR 3 NP_001764.1

View Full Product Details